Gene Transcripts

Gene Symbol
Gjb3
Entrez Gene ID
29585
Gene Name
gap junction protein, beta 3
Gene Aliases
Chromosome Location
Chr. 5: 145390590 - 145397279 on Build Rnor_6.0
UniGene ID
Rn.162823
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019240.1 NP_062113.1 1 341
XM_006238845.2 XP_006238907.1 2 1651

View Full Product Details