Gene Transcripts

Gene Symbol
Gja5
Entrez Gene ID
50563
Gene Name
gap junction protein, alpha 5
Gene Aliases
Cx40
Chromosome Location
Chr. 2: 199162745 - 199184942 on Build Rnor_6.0
UniGene ID
Rn.88300
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019280.1 NP_062153.1 2 1319
GenBank mRNA AF021806.1 - 2 1319
M83092.1 - 1 1229

View Full Product Details