Gene Transcripts
- Gene Symbol
- Sparc
- Entrez Gene ID
- 24791
- Gene Name
- secreted protein, acidic, cysteine-rich (osteonectin)
- Gene Aliases
- Chromosome Location
- Chr. 10: 40742390 - 40764222 on Build Rnor_6.0
- UniGene ID
- Rn.234316
- Species
- Rattus norvegicus
View Full Product Details