Gene Transcripts

Gene Symbol
Slc4a2
Entrez Gene ID
24780
Gene Name
solute carrier family 4 (anion exchanger), member 2
Gene Aliases
Ae2, Aep2
Chromosome Location
Chr. 4: 7264678 - 7282734 on Build Rnor_6.0
UniGene ID
Rn.9860
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017048.2 NP_058744.1 4 604
XM_006235874.2 XP_006235936.1 4 1061
XM_006235875.2 XP_006235937.1 4 782
XM_006235876.2 XP_006235938.1 4 844
XM_006235877.2 XP_006235939.1 3 987
XM_006235878.2 XP_006235940.1 3 906
GenBank mRNA AJ288902.1 - 3 399
J05166.1 - 4 608

View Full Product Details