Gene Transcripts

Gene Symbol
Parm1
Entrez Gene ID
286894
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
CIPAR-1, Cipar1, PARM-1
Chromosome Location
Chr. 14: 17972914 - 18076870 on Build Rnor_6.0
UniGene ID
Rn.127830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173114.1 NP_775137.1 2 257
GenBank mRNA AJ010750.1 - 2 279
BC060539.1 - 2 283 6890269
FQ219991.1 - 2 280

View Full Product Details