Gene Transcripts

Gene Symbol
Smptb
Entrez Gene ID
302879
Gene Name
polypyrimidine tract-binding protein
Gene Aliases
Chromosome Location
Chr. X: 17484839 - 17486721 on Build Rnor_6.0
UniGene ID
Rn.97598
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182818.1 NP_877970.1 1 277
GenBank mRNA AY223520.1 - 1 277

View Full Product Details