Gene Transcripts

Gene Symbol
C19orf53
Entrez Gene ID
28974
Gene Name
chromosome 19 open reading frame 53
Gene Aliases
HSPC023, LYDG10
Chromosome Location
Chr. 19: 13774443 - 13778773 on Build GRCh38
UniGene ID
Hs.231616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014047.2 NP_054766.1 3 571
GenBank mRNA AF078852.1 - 3 545
AF086483.1 - 3 543
BC015465.2 - 3 546 3914049

View Full Product Details