Gene Transcripts

Gene Symbol
HS3ST4
Entrez Gene ID
9951
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 4
Gene Aliases
3-OST-4, 30ST4, 3OST4, h3-OST-4
Chromosome Location
Chr. 16: 25692026 - 26137688 on Build GRCh38
UniGene ID
Hs.655275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006040.2 NP_006031.2 2 2286
GenBank mRNA AF105378.2 - 2 2284
AL834283.1 - 2 1533

View Full Product Details