Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012351.2 | NP_036483.2 | 1 | 199 | |
XM_006711271.2 | XP_006711334.1 | 2 | 211 | ||
XM_017001015.1 | XP_016856504.1 | 2 | 504 | ||
GenBank mRNA | BC069150.1 | - | 1 | 162 | |
BC112290.1 | - | 1 | 211 | 8327758 | |
BC112292.1 | - | 1 | 199 |