Gene Transcripts

Gene Symbol
OR52K2
Entrez Gene ID
119774
Gene Name
olfactory receptor family 52 subfamily K member 2
Gene Aliases
OR11-7
Chromosome Location
Chr. 11: 4449295 - 4450361 on Build GRCh38
UniGene ID
Hs.553568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005172.2 NP_001005172.2 1 443
GenBank mRNA BC137247.1 - 1 443
BC137248.1 - 1 443 9021245

View Full Product Details