Gene Transcripts

Gene Symbol
OR1M1
Entrez Gene ID
125963
Gene Name
olfactory receptor family 1 subfamily M member 1
Gene Aliases
OR19-5, OR19-6
Chromosome Location
Chr. 19: 9093245 - 9094186 on Build GRCh38
UniGene ID
Hs.553577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004456.1 NP_001004456.1 1 577
GenBank mRNA BC137551.1 - 1 643
BC137555.1 - 1 643 9021555

View Full Product Details