Gene Transcripts

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
Chromosome Location
Chr. 8: 11676919 - 11760002 on Build GRCh38
UniGene ID
Hs.243987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308093.1 NP_001295022.1 2 1095
NM_001308094.1 NP_001295023.1 3 435
NM_002052.4 NP_002043.2 3 1246
XM_005272385.4 XP_005272442.1 4 1318
XM_005272386.1 XP_005272443.1 4 1506
XM_006716248.1 XP_006716311.1 4 1427
XM_011543817.2 XP_011542119.1 4 1497
XM_011543818.2 XP_011542120.1 3 1635
XM_017013312.1 XP_016868801.1 3 1951
GenBank mRNA AK097060.1 - 3 395
AY740706.1 - 2 638
BC101580.1 - 2 715 8069086
BC105108.1 - 2 713 8144111
BC143434.1 - 2 715 9051942
BC143479.1 - 2 718 9051987
D78260.1 - 2 927
DA900813.1 - 2 176
L34357.1 - 2 875

View Full Product Details