Gene Transcripts

Gene Symbol
GRASP
Entrez Gene ID
160622
Gene Name
general receptor for phosphoinositides 1 associated scaffold protein
Gene Aliases
TAMALIN
Chromosome Location
Chr. 12: 52006945 - 52015889 on Build GRCh38
UniGene ID
Hs.407202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271856.1 NP_001258785.1 2 101
NM_181711.3 NP_859062.1 3 401
XM_005268691.3 XP_005268748.1 3 1569
XM_011537996.1 XP_011536298.1 2 1347
GenBank mRNA BC035500.2 - 2 72
BC052967.1 - 2 85
BC053666.1 - 3 382 6151168
HY035281.1 - 2 101
HY052304.1 - 3 402

View Full Product Details