Gene Transcripts

Gene Symbol
OR6V1
Entrez Gene ID
346517
Gene Name
olfactory receptor family 6 subfamily V member 1
Gene Aliases
GPR138
Chromosome Location
Chr. 7: 143052341 - 143053282 on Build GRCh38
UniGene ID
Hs.554721
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001667.1 NP_001001667.1 1 241
GenBank mRNA BC137044.1 - 1 262

View Full Product Details