Gene Transcripts
- Gene Symbol
- FAM27E5
- Entrez Gene ID
- 284123
- Gene Name
- family with sequence similarity E5
- Gene Aliases
- FAM27L
- Chromosome Location
- Chr. 17: 22298764 - 22299893 on Build GRCh38
- UniGene ID
- Hs.348578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028336.1 |
|
2 |
300 |
|
GenBank mRNA |
BC031617.1 |
- |
2 |
300 |
|
View Full Product Details