Gene Transcripts

Gene Symbol
Tmem98
Entrez Gene ID
303356
Gene Name
transmembrane protein 98
Gene Aliases
Chromosome Location
Chr. 10: 68173380 - 68184289 on Build Rnor_6.0
UniGene ID
Rn.24570
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007672.1 NP_001007673.1 3 280
GenBank mRNA BC078932.1 - 3 280
FQ214419.1 - 3 299

View Full Product Details