Gene Transcripts

Gene Symbol
COA5
Entrez Gene ID
493753
Gene Name
cytochrome c oxidase assembly factor 5
Gene Aliases
6330578E17Rik, C2orf64, CEMCOX3, Pet191
Chromosome Location
Chr. 2: 98599323 - 98608518 on Build GRCh38
UniGene ID
Hs.596537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008215.2 NP_001008216.1 3 557
GenBank mRNA AK131034.1 - 3 554
BC047722.1 - 3 531 5748383

View Full Product Details