Gene Transcripts

Gene Symbol
Prim1
Entrez Gene ID
246327
Gene Name
primase, DNA, polypeptide 1
Gene Aliases
Chromosome Location
Chr. 7: 2435553 - 2451169 on Build Rnor_6.0
UniGene ID
Rn.92497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ011608.1 - 12 1175
BC088272.1 - 12 1182 7379940
FQ231746.1 - 5 381

View Full Product Details