Gene Transcripts

Gene Symbol
Adgrl1
Entrez Gene ID
330814
Gene Name
adhesion G protein-coupled receptor L1
Gene Aliases
2900070I05Rik, AI182398, CLIBA, Lec2, Lphn1, mKIAA0821
Chromosome Location
Chr. 8: 83900098 - 83941954 on Build GRCm38
UniGene ID
Mm.260733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181039.2 NP_851382.2 5 1702
GenBank mRNA AK147455.1 - 5 1696
AK147519.1 - 3 609
BC085138.1 - 6 1103

View Full Product Details