Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002821.1 | NP_001002821.1 | 935 | ||
XM_008772742.1 | XP_008770964.1 | 5 | 1502 | ||
XM_008772743.1 | XP_008770965.1 | 6 | 967 | ||
XM_008772744.1 | XP_008770966.1 | 6 | 2105 | ||
GenBank mRNA | BC104719.1 | - | 1477 |