Gene Transcripts

Gene Symbol
Slc28a1
Entrez Gene ID
434203
Gene Name
solute carrier family 28 (sodium-coupled nucleoside transporter), member 1
Gene Aliases
Cnt1
Chromosome Location
Chr. 7: 81113269 - 81170416 on Build GRCm38
UniGene ID
Mm.389909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004184.3 NP_001004184.2 13 1372
GenBank mRNA BC061230.1 - 13 1372

View Full Product Details