Gene Transcripts

Gene Symbol
Kat5
Entrez Gene ID
192218
Gene Name
K(lysine) acetyltransferase 5
Gene Aliases
Htatip, Htatip1, Tip60
Chromosome Location
Chr. 1: 220967795 - 220974596 on Build Rnor_6.0
UniGene ID
Rn.6629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005872.1 NP_001005872.1 7 906
GenBank mRNA AF333984.1 - 5 559
BC083879.1 - 7 906 7130366

View Full Product Details