Gene Transcripts

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
Chromosome Location
Chr. 1: 92832729 - 92961522 on Build GRCh38
UniGene ID
Hs.180946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252273.1 NP_001239202.1 4, 5 575
GenBank mRNA BC070342.1 - 4, 5 509
DA736826.1 - 3, 4 316

View Full Product Details