Gene Transcripts

Gene Symbol
Olr94
Entrez Gene ID
405908
Gene Name
olfactory receptor 94
Gene Aliases
Chromosome Location
Chr. 1: 168470190 - 168471161 on Build Rnor_6.0
UniGene ID
Rn.141893
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001023.1 NP_001001023.1 1 283

View Full Product Details