Gene Transcripts

Gene Symbol
Acap3
Entrez Gene ID
140500
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
Centb5, Kiaa1716-hp, mKIAA1716
Chromosome Location
Chr. 4: 155891875 - 155907251 on Build GRCm38
UniGene ID
Mm.219591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207223.1 NP_997106.1 9 1200
GenBank mRNA AK145262.1 - 3 154
AK147505.1 - 9 1254
AK159055.1 - 6 833
BC025556.1 - 9 774
BC067016.1 - 9 1200 6827624
BC098196.1 - 9 776 6847993

View Full Product Details