Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001184985.1 | NP_001171914.1 | 9 | 3042 | |
NM_213655.4 | NP_998820.3 | 10 | 3297 | ||
XM_011520997.2 | XP_011519299.1 | 9 | 3413 | ||
XM_011520998.2 | XP_011519300.1 | 9 | 3367 | ||
XM_011520999.2 | XP_011519301.1 | 9 | 3367 | ||
XM_011521000.2 | XP_011519302.1 | 9 | 3367 | ||
XM_011521001.2 | XP_011519303.1 | 9 | 3367 | ||
XM_011521002.2 | XP_011519304.1 | 9 | 3367 | ||
XM_011521003.2 | XP_011519305.1 | 9 | 3367 | ||
XM_011521004.2 | XP_011519306.1 | 9 | 3367 | ||
XM_011521005.2 | XP_011519307.1 | 6 | 1851 | ||
GenBank mRNA | BC130467.1 | - | 1 | 332 | |
BC130469.1 | - | 1 | 332 | 40146500 | |
BJ997509.1 | - | 1 | 422 | ||
JQ358908.1 | - | 9 | 2399 |