Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 4 | 403 | |
NM_015386.2 | NP_056201.2 | 4 | 403 | ||
XM_011522981.2 | XP_011521283.1 | 5 | 563 | ||
GenBank mRNA | AB088369.1 | - | 4 | 380 | |
AK022874.1 | - | 4 | 383 | ||
AK297557.1 | - | 5 | 346 | ||
BC027726.2 | - | 4 | 377 | ||
BC072438.1 | - | 4 | 383 | 6011802 | |
BC128144.1 | - | 4 | 380 | ||
BC128145.1 | - | 4 | 380 | ||
BP282697.1 | - | 4 | 367 | ||
BX648010.1 | - | 3 | 305 |