Gene Transcripts

Gene Symbol
TRIM67
Entrez Gene ID
440730
Gene Name
tripartite motif containing 67
Gene Aliases
TNL
Chromosome Location
Chr. 1: 231162928 - 231221568 on Build GRCh38
UniGene ID
Hs.655089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004342.3 NP_001004342.3 5 1567
NM_001300889.1 NP_001287818.1 7 1381
XM_017001323.1 XP_016856812.1 5 1567
GenBank mRNA AK126782.1 - 5 1567
AY253917.1 - 7 1339

View Full Product Details