Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143775.1 | NP_001137247.1 | 5 | 815 | |
NM_015343.4 | NP_056158.2 | 6 | 882 | ||
GenBank mRNA | AJ011916.1 | - | 5 | 465 | |
AY364239.1 | - | 5 | 435 | ||
BC009295.2 | - | 6 | 881 | 4123279 |