Gene Transcripts
- Gene Symbol
- COPS7A
- Entrez Gene ID
- 50813
- Gene Name
- COP9 signalosome subunit 7A
- Gene Aliases
- CSN7, CSN7A, SGN7a
- Chromosome Location
- Chr. 12: 6723984 - 6731875 on Build GRCh38
- UniGene ID
- Hs.530823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164093.1 |
NP_001157565.1
|
5, 6 |
1018 |
|
NM_001164094.1 |
NP_001157566.1
|
6, 7 |
883 |
|
NM_001164095.2 |
NP_001157567.1
|
6, 7 |
954 |
|
NM_016319.3 |
NP_057403.1
|
6, 7 |
856 |
|
XM_005253694.2 |
XP_005253751.1
|
6, 7 |
781 |
|
GenBank mRNA |
AB014764.1 |
- |
5, 6 |
644 |
|
AB033603.1 |
- |
6, 7 |
721 |
|
AF193844.1 |
- |
5, 6 |
644 |
|
AF210052.1 |
- |
6, 7 |
785 |
|
AK021721.1 |
- |
6, 7 |
821 |
|
AK022488.1 |
- |
6, 7 |
763 |
|
AK128792.1 |
- |
4, 5 |
553 |
|
AK292621.1 |
- |
5, 6 |
1014 |
|
BC011789.2 |
- |
6, 7 |
744 |
4025790 |
BC093015.1 |
- |
6, 7 |
795 |
30344094 |
View Full Product Details