Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286371.1 | NP_001273300.1 | 5 | 578 | |
NM_001286372.1 | NP_001273301.1 | 4 | 483 | ||
NM_016485.4 | NP_057569.2 | 5 | 578 | ||
GenBank mRNA | AF060225.1 | - | 5 | 468 | |
AF141341.1 | - | 5 | 455 | ||
AF151062.1 | - | 5 | 478 | ||
AF271994.1 | - | 5 | 467 | ||
AJ420507.1 | - | 5 | 473 | ||
AK000051.1 | - | 5 | 467 | ||
AK130738.1 | - | 5 | 470 | ||
AK301376.1 | - | 4 | 386 | ||
AL136684.1 | - | 5 | 471 | ||
AY007093.1 | - | 4 | 282 | ||
BC005937.1 | - | 5 | 463 | 4081455 | |
BC006989.1 | - | 5 | 477 | 3961637 | |
BC022536.1 | - | 5 | 477 | 4817057 | |
BM806661.1 | - | 5 | 378 | ||
CR457207.1 | - | 5 | 452 |