Gene Transcripts

Gene Symbol
CPLX4
Entrez Gene ID
339302
Gene Name
complexin 4
Gene Aliases
CPX-IV, CPXIV
Chromosome Location
Chr. 18: 59295402 - 59318649 on Build GRCh38
UniGene ID
Hs.449884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181654.3 NP_857637.1 3 1425
GenBank mRNA AB593158.1 - 3 1406
AK123184.1 - 3 1425
BX640655.1 - 3 1404
BX648518.1 - 3 1408

View Full Product Details