Gene Transcripts

Gene Symbol
Glipr1
Entrez Gene ID
299783
Gene Name
GLI pathogenesis-related 1
Gene Aliases
Chromosome Location
Chr. 7: 54767828 - 54778823 on Build Rnor_6.0
UniGene ID
Rn.65764
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011987.1 NP_001011987.1 1 256
GenBank mRNA BC089858.1 - 1 256
FQ219892.1 - 1 261
FQ220342.1 - 1 262
FQ223414.1 - 1 261
FQ226490.1 - 1 277
FQ227021.1 - 1 276
FQ229493.1 - 1 283
FQ229810.1 - 1 322
FQ230706.1 - 1 284

View Full Product Details