Gene Transcripts

Gene Symbol
Rogdi
Entrez Gene ID
287061
Gene Name
rogdi homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 10: 10761477 - 10766096 on Build Rnor_6.0
UniGene ID
Rn.995
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024864.1 NP_001020035.1 6 565
GenBank mRNA BC083789.1 - 4 996
BC098001.1 - 6 565 7453517

View Full Product Details