Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004070.2 | NP_001004070.1 | 2, 3 | 216 | |
NM_001165880.1 | NP_001159352.1 | 3, 4 | 364 | ||
XM_006255977.2 | XP_006256039.1 | 3, 4 | 1085 | ||
XM_006255978.2 | XP_006256040.1 | 4, 5 | 971 | ||
XM_006255979.2 | XP_006256041.1 | 4, 5 | 467 | ||
GenBank mRNA | BC127463.1 | - | 3, 4 | 364 |