Gene Transcripts

Gene Symbol
OR5K1
Entrez Gene ID
26339
Gene Name
olfactory receptor family 5 subfamily K member 1
Gene Aliases
HSHTPCRX10, HTPCRX10, OR3-8
Chromosome Location
Chr. 3: 98469480 - 98470576 on Build GRCh38
UniGene ID
Hs.621533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004736.3 NP_001004736.2 1 985
GenBank mRNA BC136918.1 - 1 985
BC136919.1 - 1 985 9020910

View Full Product Details