Gene Transcripts

Gene Symbol
OR51A7
Entrez Gene ID
119687
Gene Name
olfactory receptor family 51 subfamily A member 7
Gene Aliases
OR11-27
Chromosome Location
Chr. 11: 4907370 - 4908308 on Build GRCh38
UniGene ID
Hs.553560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004749.1 NP_001004749.1 1 888
GenBank mRNA BC137062.1 - 1 925
BC137063.1 - 1 925 9021060

View Full Product Details