Gene Transcripts

Gene Symbol
Upf3a
Entrez Gene ID
361176
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Gene Aliases
Chromosome Location
Chr. 16: 81521084 - 81532512 on Build Rnor_6.0
UniGene ID
Rn.56460
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012159.1 NP_001012159.1 3 363
GenBank mRNA BC088463.1 - 3 363

View Full Product Details