Gene Transcripts

Gene Symbol
Hsp90b1
Entrez Gene ID
362862
Gene Name
heat shock protein 90, beta, member 1
Gene Aliases
Grp94, Tra1
Chromosome Location
Chr. 7: 27226570 - 27240533 on Build Rnor_6.0
UniGene ID
Rn.101146
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012197.2 NP_001012197.2 12 1680
GenBank mRNA BC081917.1 - 12 1680
FQ229309.1 - 1 94

View Full Product Details