Gene Transcripts

Gene Symbol
Tmc5
Entrez Gene ID
365360
Gene Name
transmembrane channel-like 5
Gene Aliases
Chromosome Location
Chr. 1: 188288738 - 188337310 on Build Rnor_6.0
UniGene ID
Rn.145296
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012216.1 NP_001012216.1 6 1186
GenBank mRNA BC088406.1 - 6 1186

View Full Product Details