Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013234.1 | NP_001013252.1 | 8 | 1293 | |
XM_008772414.1 | XP_008770636.1 | 8 | 823 | ||
XM_008772415.1 | XP_008770637.1 | 6 | 607 | ||
XM_008772416.1 | XP_008770638.1 | 6 | 603 | ||
GenBank mRNA | BC086345.1 | - | 8 | 823 | |
BC086539.1 | - | 8 | 1293 | 7114943 | |
FQ227287.1 | - | 6 | 652 |