Gene Transcripts

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
Chromosome Location
Chr. 1: 152348735 - 152360028 on Build GRCh38
UniGene ID
Hs.156124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014342.2 NP_001014364.1 3 7552
XM_011509531.2 XP_011507833.1 4 7118
GenBank mRNA AY827490.1 - 3 7553

View Full Product Details