Gene Transcripts

Gene Symbol
PRR18
Entrez Gene ID
285800
Gene Name
proline rich 18
Gene Aliases
Chromosome Location
Chr. 6: 166305680 - 166308383 on Build GRCh38
UniGene ID
Hs.731862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175922.3 NP_787118.2 1 1722
GenBank mRNA BC034775.1 - 1 1722

View Full Product Details