Gene Transcripts

Gene Symbol
Scrn1
Entrez Gene ID
502776
Gene Name
secernin 1
Gene Aliases
Chromosome Location
Chr. 4: 84678978 - 84740988 on Build Rnor_6.0
UniGene ID
Rn.154317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025063.1 NP_001020234.1 6 931
XM_006236512.2 XP_006236574.1 6 919
XM_008762916.1 XP_008761138.1 6 783
GenBank mRNA BC079152.1 - 6 931

View Full Product Details