Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001720.1 | NP_001001720.1 | 2 | 323 | |
XM_008766765.1 | XP_008764987.1 | 3 | 423 | ||
XM_008766766.1 | XP_008764988.1 | 2 | 477 | ||
GenBank mRNA | AY521229.1 | - | 2 | 323 |