Gene Transcripts

Gene Symbol
Ccbl2
Entrez Gene ID
541589
Gene Name
cysteine conjugate-beta lyase 2
Gene Aliases
Kat3
Chromosome Location
Chr. 2: 248644473 - 248690289 on Build Rnor_6.0
UniGene ID
Rn.28263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015037.1 NP_001015037.1 1647
GenBank mRNA AY955395.1 - 1647

View Full Product Details