Gene Transcripts

Gene Symbol
Ddi2
Entrez Gene ID
68817
Gene Name
DNA-damage inducible protein 2
Gene Aliases
1110056G13Rik, 1700027M01Rik, 9130022E05Rik, AI604911, AU040698
Chromosome Location
Chr. 4: 141683563 - 141723419 on Build GRCm38
UniGene ID
Mm.393721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017966.2 NP_001017966.1 8 1389
GenBank mRNA BC021415.1 - 8 1370
BC031491.1 - 1 143
CD550969.1 - 3 301

View Full Product Details