Gene Transcripts
- Gene Symbol
- DOCK10
- Entrez Gene ID
- 55619
- Gene Name
- dedicator of cytokinesis 10
- Gene Aliases
- DRIP2, Nbla10300, ZIZ3
- Chromosome Location
- Chr. 2: 224765090 - 225042689 on Build GRCh38
- UniGene ID
- Hs.46578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290263.1 |
NP_001277192.1
|
39, 40 |
4356 |
|
NM_014689.2 |
NP_055504.2
|
39, 40 |
4499 |
|
XM_005246673.4 |
XP_005246730.1
|
39, 40 |
4358 |
|
XM_006712617.3 |
XP_006712680.1
|
39, 40 |
4523 |
|
XM_017004441.1 |
XP_016859930.1
|
39, 40 |
4526 |
|
XM_017004442.1 |
XP_016859931.1
|
39, 40 |
4502 |
|
XM_017004443.1 |
XP_016859932.1
|
39, 40 |
4575 |
|
XM_017004444.1 |
XP_016859933.1
|
38, 39 |
4482 |
|
XM_017004445.1 |
XP_016859934.1
|
39, 40 |
4356 |
|
XM_017004446.1 |
XP_016859935.1
|
38, 39 |
4380 |
|
XM_017004447.1 |
XP_016859936.1
|
39, 40 |
4498 |
|
GenBank mRNA |
AK293847.1 |
- |
5, 6 |
425 |
|
AK293879.1 |
- |
10, 11 |
905 |
|
CR749492.1 |
- |
14, 15 |
1344 |
|
EU236710.2 |
- |
39, 40 |
4353 |
|
View Full Product Details