Gene Transcripts

Gene Symbol
Plxnd1
Entrez Gene ID
67784
Gene Name
plexin D1
Gene Aliases
6230425C21Rik, b2b1863Clo, b2b553Clo
Chromosome Location
Chr. 6: 115954811 - 115995005 on Build GRCm38
UniGene ID
Mm.3085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026376.3 NP_080652.2 35, 36 5856
GenBank mRNA AK129175.1 - 35, 36 5121
AK147513.1 - 35, 36 5857
AY688678.1 - 35, 36 5816

View Full Product Details