Gene Transcripts

Gene Symbol
Ubxn4
Entrez Gene ID
304766
Gene Name
UBX domain protein 4
Gene Aliases
Ubxd2
Chromosome Location
Chr. 13: 44956963 - 44989653 on Build Rnor_6.0
UniGene ID
Rn.2022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012025.1 NP_001012025.1 13 1954
XM_006249699.2 XP_006249761.1 13 2000
GenBank mRNA BC088845.1 - 13 1954

View Full Product Details